missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals SGLT2/SLC5A2 Antibody (3G8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00006524-M01
This item is not returnable.
View return policy
Description
SGLT2/SLC5A2 Monoclonal antibody specifically detects SGLT2/SLC5A2 in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| SGLT2/SLC5A2 | |
| Monoclonal | |
| Unconjugated | |
| Low affinity sodium-glucose cotransporter, Na(+)/glucose cotransporter 2, SGLT2sodium/glucose cotransporter 2, solute carrier family 5 (sodium/glucose cotransporter), member 2, solute carrier family 5 (sodium/glucose transporter), member 2, Solute carrier family 5 member 2 | |
| SLC5A2 (NP_003032.1, 228 a.a. ~ 277 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GLFDKYLGAATSLTVSEDPAVGNISSFCYRPRPDSYHLLRHPVTGDLPWP | |
| 0.1 mg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 6524 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 3G8 | |
| In 1x PBS, pH 7.4 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction