missing translation for 'onlineSavingsMsg'
Learn More

Cytomegalovirus p65 Antibody (CMV100), Novus Biologicals™

Product Code. 18444972 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
Unit Size:
0.02mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18444972 0.1 mg 0.1mg
18467311 0.02 mg 0.02mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18444972 Supplier Novus Biologicals Supplier No. NBP2343320.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytomegalovirus p65 Monoclonal antibody specifically detects Cytomegalovirus p65 in Cytomegalovirus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytomegalovirus p65
Applications ELISA
Classification Monoclonal
Clone CMV100
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution ELISA 1-5 ug/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CMV, p65
Host Species Mouse
Immunogen Recombinant Cytomegalovirus p65 protein
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Cytomegalovirus (CMV)
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.