Learn More
Fisher Bioreagents 0.01 KG AMMONIUM PERSULPHATE TABLETS pack of 100x100mg, each tablet makes 1ml of 10% solution
Brand:  Fisher Bioreagents A1520-10G
0.01 KG AMMONIUM PERSULPHATE TABLETS pack of 100x100mg, each tablet makes 1ml of 10% solution
Product Code. 12831680