siRNA Details
| Gene Symbol: Maneal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238913.2 | XP_006238975.1 | 3 | 1482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Maneal| Gene Symbol: Maneal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238913.2 | XP_006238975.1 | 3 | 1482 | ||