siRNA Details
| Gene Symbol: DNAH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277115.1 | NP_001264044.1 | 15 | 2802 | ||
| GenBank mRNA | AJ320497.1 | - | 15 | 2802 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAH11| Gene Symbol: DNAH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277115.1 | NP_001264044.1 | 15 | 2802 | ||
| GenBank mRNA | AJ320497.1 | - | 15 | 2802 | ||