siRNA Details
| Gene Symbol: CCDC169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023568.1 | - | 1 | 1300 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX116702.1 | - | 1 | 122 | Chr. 13 - 36222008 - 36222817 [-] on Build GRCh38 | |

CCDC169
Genomic Map