siRNA Details
| Gene Symbol: Wbscr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025112.1 | NP_001020283.1 | 8 | 1332 | ||
| XM_008769132.1 | XP_008767354.1 | 8 | 1200 | |||
| GenBank mRNA | AB040671.1 | - | 8 | 1332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wbscr17| Gene Symbol: Wbscr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025112.1 | NP_001020283.1 | 8 | 1332 | ||
| XM_008769132.1 | XP_008767354.1 | 8 | 1200 | |||
| GenBank mRNA | AB040671.1 | - | 8 | 1332 | ||