siRNA Details
| Gene Symbol: LOC103693605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768923.1 | XP_008767145.1 | 3, 4 | 2067 | ||
| XM_008768924.1 | XP_008767146.1 | 2, 3 | 2031 | |||
| XM_008768925.1 | XP_008767147.1 | 3, 4 | 2067 | |||
| XM_008768926.1 | XP_008767148.1 | 3, 4 | 2067 | |||
| Gene Symbol: Car15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105901.1 | NP_001099371.1 | 3, 4 | 349 | ||
| XM_008768867.1 | XP_008767089.1 | 2, 3 | 2031 | |||
| XM_008768868.1 | XP_008767090.1 | 3, 4 | 2067 | |||
| XM_008768869.1 | XP_008767091.1 | 3, 4 | 2067 | |||

LOC103693605
Genomic Map