siRNA Details
| Gene Symbol: MEGF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001409.3 | NP_001400.3 | 23 | 3100 | ||
| XM_006710406.3 | XP_006710469.1 | 20 | 2735 | |||
| XM_011540885.1 | XP_011539187.1 | 24 | 3405 | |||
| XM_011540886.1 | XP_011539188.1 | 24 | 3405 | |||
| XM_011540887.2 | XP_011539189.1 | 19 | 2613 | |||
| XM_011540888.2 | XP_011539190.1 | 24 | 3405 | |||
| XM_017000533.1 | XP_016856022.1 | 23 | 3273 | |||
| GenBank mRNA | AB011539.2 | - | 20 | 2761 | ||
| AB231860.1 | - | 23 | 3100 | |||

MEGF6
Genomic Map