siRNA Details
| Gene Symbol: CC2D1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017721.4 | NP_060191.3 | 19 | 2331 | ||
| XM_005259972.2 | XP_005260029.1 | 19 | 2276 | |||
| XM_005259973.2 | XP_005260030.1 | 19 | 2279 | |||
| XM_005259974.2 | XP_005260031.1 | 19 | 2267 | |||
| XM_005259975.2 | XP_005260032.1 | 19 | 2264 | |||
| GenBank mRNA | AB097002.1 | - | 19 | 2273 | ||
| AF536205.1 | - | 21 | 2384 | |||
| AK000248.1 | - | 683 | ||||
| AK023399.1 | - | 12 | 1156 | |||
| AK026371.1 | - | 3 | 169 | |||
| AK315068.1 | - | 19 | 2249 | |||
| BC006556.2 | - | 14 | 1435 | |||
| BC048345.1 | - | 5 | 343 | |||
| BC064981.1 | - | 19 | 2235 | 6585236 | ||

CC2D1A
Genomic Map