siRNA Details
| Gene Symbol: TAF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005645.3 | NP_005636.1 | 4 | 276 | ||
| GenBank mRNA | AK312158.1 | - | 4 | 272 | ||
| AU280379.1 | - | 4 | 276 | |||
| AV713996.1 | - | 4 | 223 | |||
| BC121180.1 | - | 4 | 250 | 40123243 | ||
| BC121181.1 | - | 4 | 250 | 40123247 | ||
| BG675152.1 | - | 2 | 84 | |||
| X84003.1 | - | 4 | 232 | |||

TAF13
Genomic Map