siRNA Details
| Gene Symbol: Ccnjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045530.2 | NP_001038995.1 | 3 | 374 | ||
| GenBank mRNA | AK143205.1 | - | 3 | 340 | ||
| BC139344.1 | - | 2 | 135 | 8862366 | ||
| BC139345.1 | - | 2 | 135 | 8862367 | ||
| CJ066789.1 | - | 3 | 377 | |||

Ccnjl
Genomic Map