siRNA Details
| Gene Symbol: GLUD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318900.1 | NP_001305829.1 | 13 | 2491 | ||
| NM_001318901.1 | NP_001305830.1 | 16 | 2844 | |||
| NM_001318902.1 | NP_001305831.1 | 15 | 2771 | |||
| NM_001318904.1 | NP_001305833.1 | 14 | 3286 | |||
| NM_001318905.1 | NP_001305834.1 | 16 | 3412 | |||
| NM_001318906.1 | NP_001305835.1 | 14 | 3119 | |||
| NM_005271.4 | NP_005262.1 | 13 | 3059 | |||
| GenBank mRNA | AB209406.1 | - | 12 | 5359 | ||
| AK094782.1 | - | 15 | 2723 | |||
| AK122685.1 | - | 16 | 2827 | |||
| BC112946.1 | - | 13 | 2842 | 6138490 | ||
| J03248.1 | - | 13 | 2842 | |||
| M20867.1 | - | 13 | 2878 | |||
| M37154.1 | - | 13 | 2794 | |||
| X07674.1 | - | 13 | 2794 | |||
| X07769.1 | - | 13 | 2794 | |||

GLUD1
Genomic Map