Use promo code "24079" to get your promotional price.
Thermo Scientific™ Pierce™ BCA Protein Assay Kits
The Pierce BCA Protein Assay Kit is a high-precision, detergent-compatible protein assay for determination of protein concentration. Pierce BCA reagents provide accurate determination of protein concentration with most sample types encountered in protein research.
152.76 GBP valid until 2025-12-31
Use promo code "24079" to get your promotional price.
Use promo code "24079" to get your promotional price.
Code:
PN
Additional Details:
CAS Number:
7758-99-8
Weight:
0.60000kg
Transport:
UN number : 3082
Chem class : 9
Pack group : III
Disclaimers:
For Research Use Only.
Specification
| Concentration | BSA: 2 mg/mL |
|---|---|
| Content And Storage |
Sufficient For: 250 tube assays or 2500 microplate assays • BCA Reagent A, 500 mL • BCA Reagent B, 25 mL • Albumin Standard Ampules, 2 mg/mL, 10 x 1 mL Store at room temperature |
| Assay | BCA Assay |
| Detection Method | Colorimetric |
| Product Line | Pierce |
| Product Type | Protein Quantitation Assay |
| Specificity | Not Target-Specific |
| Sufficient For | 250 Tube Assays or 2500 Microplate Assays |
| For Use With (Application) | Solution-based Detection, Absorbance |
| For Use With (Equipment) | Spectrophotometer, Microplate Reader |
| Kit Contents | 10 x 1 mL glass ampules |
| Quantity | 1 Kit (500 mL) |
Technical documents
[SET BCA PROTEIN ASSAY KIT, 500ML KIT]
[Warning]
[{code=H400, sequence=1, description=Very toxic to aquatic life.}, {code=H411, sequence=2, description=Toxic to aquatic life with long lasting effects.}]
[{code=P273, sequence=51, description=Avoid release to the environment.}, {code=P391, sequence=52, description=Collect spillage.}, {code=P501, sequence=53, description=Dispose of contents/container to .}]
[{code=MIXTURE LIST, sequence=1, description=Contain: Copper (II) sulfatepentahydrate}]
[{code=GHS09, sequence=1, description=ENVIRONMENT}]
Product Identifier
- SET BCA PROTEIN ASSAY KIT, 500ML KIT
Signal Word
- Warning
Hazard Statement
- H400- Very toxic to aquatic life.
- H411- Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273- Avoid release to the environment.
- P391- Collect spillage.
- P501- Dispose of contents/container to .
Supplemental Information
- MIXTURE LIST- Contain: Copper (II) sulfatepentahydrate