235.14 GBP valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Use promo code "24140" to get your promotional price.
| Accuracy | ±1 mL |
|---|---|
| Material | Polystyrene |
| Sterility | Sterile |
| Graduated | Yes |
| Graduations | 0.5 mL |
| Length (English) | 13.5 in. |
| Quantity | 160 Pk. (4 Bags of 40) |
| Color | Violet |
| Volume (Metric) | 50 mL |
| Length (Metric) | 343 mm |
| Type | Serological Pipet |
| Disposable | Single-use |