Cross reference/Alternative names
A
| Alternative Name | Catalogue Name | Ref |
| Acacia | Gum acacia | G/1050 |
| Acetylene tetrabromide | 1,1,2,2-Tetrabromoethane | 18087 |
| Adipyl chloride | Adipoyl chloride | A/1020 |
| Adsorbents | Aluminium oxides | A/2280 |
| Adsorbents | Florisil | F/1170 |
| Adsorbents | Molecular sieves | MS/1010 |
| Alcohol, industrial | Methylated spirit, industrial | M/4400 |
| Alizarin complexone | 3-Aminomethylalizarin-N, N-diacetic acid | A/2930 |
| Alizarin fl uorine blue | 3-Aminomethylalizarin-N, N-diacetic acid | A/2930 |
| Alum, ammonium | Aluminium ammonium sulfate | A/1840 |
| Alum, chrome | Chromium potassium sulfate | C/5720 |
| Alum, iron | Ammonium iron (III) sulfate | A/4720 |
| Alum, potassium | Aluminium potassium sulfate | A/2400 |
| Alumina | Aluminium oxide | A/2280 |
| 1-Amino butane | n-Butylamine | B/5260 |
| Aminoacetic acid | Glycine | G/0800 |
| 4-Aminoantipyrine | 4-Aminophenazone | A/3040 |
| 4-Aminobenzene sulfonamide | Sulfanilamide | S/8660 |
| 4-Aminobenzenesulfonic acid | Sulfanilic acid | S/8680 |
| 2-Aminobenzoic acid | Anthranilic acid | A/7680 |
| 4-Amino-N,N-dimethylaniline sulfate | N, N-Dimethyl-p-phenylenediamine sulfate | D/4016 |
| Aminoethane | Ethylamine | E/1000 |
| 2-Aminoethanol | Ethanolamine | E/0700 |
| 4-Amino-3-hydroxynaphthalene-1-sulfonic acid | 1-Amino-2-naphthol-4-sulfonic acid | A/3002 |
| 8-Aminonaphthalene-2-sulfonic acid | 1-Naphthylamine-7-sulfonic acid | N/0910 |
| Ammonia-ammonium chloride buffer | Buffer solution, ammonia-ammonium chloride | J/2500 |
| Ammonium alum | Aluminium ammonium sulfate | A/1840 |
| Ammonium bicarbonate | Ammonium hydrogen carbonate | A/5120 |
| Ammonium bichromate | Ammonium dichromate | A/4400 |
| Ammonium bifl uoride | Ammonium hydrogen difl uoride | A/5240 |
| Ammonium bisulfate | Ammonium hydrogen sulfate | A/5400 |
| Ammonium citrate | di-Ammonium hydrogen citrate | A/5200 |
| Ammonium ferric salts | Ammonium iron (III) sulfate | A/4760 |
| Ammonium ferrous salts | Ammonium iron (II) sulfate | A/4880 |
| Ammonium fl uoride, acid | Ammonium hydrogen difl uoride | A/5280 |
| Ammonium hydroxide | Ammonia solutions | A/3240 |
| Ammonium methanoate | Ammonium formate | A/5040 |
| Ammonium phosphate dibasic | di-Ammonium hydrogen orthophosphate | A/5320 |
| Ammonium phosphate monobasic | Ammonium dihydrogen orthophosphate | A/4520 |
| Ammonium sulfocyanide | Ammonium thiocyanate | A/6640 |
| Ammonium tetramethylenedithio-carbamate | Ammonium pyrrolidine dithiocarbamate | A/6220 |
| Ammonium vanadate | Ammonium metavanadate | A/5680 |
| tert-Amyl alcohol | 2-Methylbutan-2-ol | M/4480 |
| n-Amyl alcohol | Pentan-1-ol | P/1040 |
| sec-Amyl alcohol | Pentan-2-ol | P/1080 |
| Anhydrone | Magnesium perchlorate | M/0950 |
| Anisic aldehyde | p-Anisaldehyde | A/7490 |
| Antifoaming agent | Silicone antifoaming agent | S/0820 |
| APDC | Ammonium pyrrolidine dithiocarbamate | A/6220 |
| Arsenic (III) oxide | Arsenic trioxide | A/8440 |
| Arsenous oxide | Arsenic trioxide | A/8440 |
| Aspirin | Acetylsalicylic acid | A/0960 |
B
| Alternative Name | Catalogue Name | Ref |
| Beeswax | Wax, beeswax | W/0150 |
| BHA | Butylated hydroxyanisole | B/5270 |
| BHT | 2,6-Di-tert-butyl-p-cresol | D/1345 |
| BIS or Bis Acrylamide | Methylene bis acrylamide | M/P210 |
| N,N-Bis(2-hydroxyethyl)-2-amino-ethansulfonic acid | BES biological buffer | BPE501 |
| Bis-(3,5,5-trimethylhexyl) phthalate | Dinonyl phthalate | D/4472 |
| Bis-Tris | Bis(2-hydroxyethyl)-imino-tris-(hydroxymethyl)-methane | B/3010 |
| Bleaching powder | Calcium hypochlorite | C/1720 |
| Borax | di-Sodium tetraborate | S/7000 |
| Borax glass | di-Sodium tetraborate anhydrous | S/6980 |
| Boric acid solution | Kjeldahl reagents | K/0200 |
| Brilliant Blue G-250 | Coomassie blue | BPE100 |
| Brilliant Blue R-250 | Coomassie blue | BPE101 |
| BSA | Bis (trimethylsilyl) acetamide or Bovine serum albumin | B/3640, A/1278 |
| BSTFA | Bis (trimethylsilyl) trifl uoroacetamide | B/3647 |
| Butanedioic acid | Succinic acid | S/8520 |
| Butane-2,3-dione | Diacetyl | D/0250 |
| Butan-2-ol | sec-Butyl alcohol | B/5200 |
| n-Butanol | Butan-1-ol | B/4800 |
| Butan-2-one | Ethyl methyl ketone | E/1450 |
| n-Butyl alcohol | Butan-1-ol | B/4800 |
| n-Butyl bromide | 1-Bromobutane | B/4310 |
| Butyl carbitol | Butyl digol | B/5300 |
| Butyl cellosolve | 2-n-Butoxyethanol | B/4900 |
| n-Butyl chloride | 1-Chlorobutane | C/4750 |
| tert-Butyl chloride | 2-Chloro-2-methyl propane | C/4990 |
| n-Butyl ether | di-N-butyl ether | D/1350 |
| n-Butyl iodide | 1-Iodobutane | I/0697 |
| tert-Butyl methyl ether | Methyl-tert-butyl ether | M/4494 |
| iso-Butyl methyl ketone | 4-Methylpentan-2-one | M/5078 |
| iso-Butyl methyl ketone for A.A.S, | 4-Methylpentan-2-one | M/5080 |
| n-Butyl phosphate | tri-n-Butyl phosphate | 16993 |
| Butyl phthalate | di-n-butyl phthalate | D/1500 |
| Butylated hydroxytoluene | 2,6-Di-tert-butyl-p-cresol | D/1345 |
| Butyl ethanoate | n-Butyl acetate | B/4950 |
C
| Alternative Name | Catalogue Name | Ref |
| Calcein | Fluorescein di-(methylimino diacetic acid), sodium salt | F/1250 |
| Calcium hydroxide solution | Lime water | J/5000 |
| Calcium pantothenate | (+)-Pantothenic acid calcium salt | P/0295 |
| Calcium phosphate, precipitated | tri-Calcium di-orthophosphate | C/1960 |
| Calcon | Eriochrome blue-black R | E/0200 |
| Calmagite | 1-(1-Hydroxy-4-methyl-2-phenylazo)-2-naphthol-4-sulfonic acid | H/2350 |
| Capryl alcohol | Octan-2-ol | O/0100 |
| Caprylic acid | n-Octanoic acid | O/0030 |
| Carbamide | Urea | U/0450 |
| Carbitol | Ethyl digol | E/1300 |
| 3-Carboxy-4-hydroxybenzene sulfonic acid | 5-Sulfosalicylic acid | S/8850 |
| Carnauba wax | Wax, carnauba | W/0255 |
| Catechol violet | Catecholsulfonaphthalein | C/3360 |
| Celite(R)-521 | Kieselguhr | K/0100 |
| Ceramic fi bre wool | Triton Kaowool(R) | T/3740 |
| Ceresin | Wax, ceresin, white | W/0300 |
| Ceric (IV) ammonium salts | Ammonium cerium nitrate | A/3720 |
| Ceric salts | Cerium (IV) sulfate | C/3720 |
| Cerous salts | Cerium (III) nitrate | C/3780 |
| Cesium chloride | Caesium chloride | C/0640 |
| Cetrimide | Tetradecyltrimethyl ammonium bromide | T/0611 |
| Chalk, French | Talc | T/0050 |
| Chloroplatinic acid | Hexachloroplatinic acid | H/0262 |
| Chlorotrimethylsilane | Trimethylchlorosilane | T/3520 |
| Chrome alum | Chromium (III) potassium sulfate | C/5720 |
| Chromic anhydride | Chromium (VI) oxide | C/5840 |
| Chromium trioxide | Chromium (VI) oxide | C/5920 |
| Cinnamic aldehyde | Cinnamaldehyde | C/6098 |
| Cleaning agents | Lotoxane(R) | L/2550 |
| Cleland's reagent | Dithiothreitol | D/P351 |
| Cleve's acid | 1-Naphthylamine-7-sulfonic acid | N/0910 |
| Completely Denatured Alcohol (CDA) | Methylated spirits | M/4350 |
| Copper ammonium salts | Ammonium copper (II) chloride | A/4240 |
| Cotton wool | Wool, cotton | CTC-230 |
| o-Cresolphthalein complexone | o-Cresolphthaleindi-(methyliminidi-acetic acid) | C/7440 |
| m-Cresolsulfonphthalein | m-Cresol purple | C/7450L |
| o-Cresolsulfonphthalein | Cresol red | C/7460 |
| Crystal violet | Methyl violet technical | M/5350 |
| Cupric ammonium salts | Ammonium copper (II) chloride | A/4240 |
D
| Alternative Name | Catalogue Name | Ref |
| Decahydronaphthalene | Dekalin | D/0050 |
| Dextrose | D-Glucose | G/0450 |
| Dichlorodimethylsilane | Dimethyldichlorosilane | D/3820 |
| Diethylbarbituric acid | Barbitone | B/0052 |
| Diethylene dioxide | Dioxan | D/4500 |
| Diethylene glycol | Digol | D/2950 |
| Diethylene glycol dimethyl ether | Dimethyl digol | D/3830 |
| Diethylene glycol monobutyl ether | Butyl digol | B/5300 |
| Diethylene glycol monoethyl ether | Ethyl digol | E/1300 |
| Diethylene glycol monomethyl ether | Methyl diglycol | M/4720 |
| Diglyme | Dimethyl digol | D/3830 |
| 1,4-Dihydroxybenzene | Quinol | Q/0150 |
| 2,3-Dihydroxybutanedioic acid | (+)-Tartaric acid | T/0250 |
| 1,8-Dihydroxynaphthalene-3,6-disulfonic acid sodium salt | Chromotropic acid sodium salt | C/6002 |
| 3,3'-(3,3'-Dimethoxy-4,4'- | ||
| biphenylene) bis (2,5-diphenyl 2H | Blue tetrazolium | B/3660 |
| tetrazolium chloride) | ||
| 4-Dimethylaminoazobenzene-4-sulfonic acid sodium salt | Methyl orange | M/5000 |
| Dimethylbenzene | Xylene | X/0100 |
| 5,5-Dimethyl-1,3 cyclohexanedione | Dimedone | D/3220 |
| 1,5-Diphenylcarbohydrazide | 1,5-Diphenylcarbazide | D/4750 |
| 1,5-Diphenylthiocarbazone | Dithizone | D/5250 |
| DMF | Dimethylformamide | D/3840 |
| Dodecanoic acid | Lauric acid | L/0560 |
| Dodecyl sodium sulfate | Sodium dodecyl sulfate | S/5200 |
| DTT | Dithiothreitol | D/P351 |
E
| Alternative Name | Catalogue Name | Ref |
| EGTA | 1,2-Di(2-aminoethoxy)ethane-N, N,N'N'-tetra-acetic acid | D/0755 |
| Ehrlich's reagent | 4-Dimethylaminobenzaldehyde | D/3550 |
| Eosin | Tetrabromo (R) fl uorescein | T/0470 |
| Eosin-methylene blue | Leishman's stain | L/1815 |
| Epsom salts | Magnesium sulfate | M/1000 |
| Ethanal | Acetaldehyde | A/0080 |
| Ethanamide | Acetamide | A/0160 |
| Ethanoic acid | Acetic acid glacial | A/0360 |
| Ethanoic acid | Acetic acid solution | J/0549 |
| Ethanoic anhydride | Acetic anhydride | A/0440 |
| ethanoyl chloride | Acetyl chloride | A/0880 |
| Ether | Diethyl ether | D/2400 |
| Ether, petroleum | Petroleum ether | P/1360 |
| Ethyl alcohol | Ethanol | E/0400 |
| Ethyl bromide | Bromoethane | B/4350 |
| Ethylene chlorhydrin | 2-Chloroethanol | C/4880 |
| Ethylene chloride | 1,2-Dichloroethane | D/1750 |
| Ethylenediamine and derivatives | 1,2-Diaminoethane anhydrous | D/0370 |
| Ethylene dibromide | 1,2-Dibromoethane | D/1250 |
| Ethylene dichloride | 1,2-Dichloroethane | D/1750 |
| Ethylene glycol | Ethanediol | E/0350 |
| Ethylene glycol bis(aminoethylether)-tetra-acetic acid | 1,2-Di(2-aminoethoxy)ethane-N, N,N'N'-tetra-acetic acid | D/0755 |
| Ethylene glycol dimethyl ether | 1,2-Dimethoxyethane | D/3280 |
| Ethylene glycol monobutyl ether | 2-n-Butoxyethanol | B/4900 |
| Ethylene glycol monoethyl ether | 2-Ethoxyethanol | E/0750 |
| Ethylene glycol monoethyl ether, acetic ester | 2-Ethoxyethyl acetate | E/0800 |
| Ethylene glycol monomethyl ether | 2-Methoxyethanol | M/4100 |
| Ethyl ethanoate | Ethyl acetate | E/0850 |
| Ethyl iodide | Iodoethane | I/0700 |
F
| Alternative Name | Catalogue Name | Ref |
| Ferric alum | Ammonium iron (III) sulfate | A/4720 |
| Ferric ammonium salts | Ammonium iron (III) sulfate | A/4720 |
| Ferric salts | Iron (III) chloride | I/1020 |
| Ferroin | 1,10-Phenanthroline ferrous complex | P/2220 |
| Ferrous ammonium salts | Ammonium iron (II) sulfate solution | A/4840 |
| Filter agent | Kieselguhr | K/0100 |
| Fluorescein complexone | Fluorescein di-(methylimino diacetic acid) | F/1250 |
| Folin's amino acid reagent | 1,2-Napthoquinone-4-sulfonic acid sodium salt | N/0230 |
| Formal saline | Formaldehyde solution | F/1451 |
| Formalin | Formaldehyde solution | F/1451 |
| Formdimethylamide | N,N-Dimethylformamide | D/3840 |
| French chalk | Talc | T/0050 |
| Full range indicator | Universal indicator | U/0020 |
| Furfural | Furfuraldehyde | F/2080 |
G
| Alternative Name | Catalogue Name | Ref |
| Gallic acid | 3,4,5-Trihydroxybenzoic acid | T/3300 |
| Gas analysis solution | Potassium pyrogallate | J/7000 |
| Gentian violet | Methyl violet 2B | M/5345 |
| Gluconic acid lactone | 1,5-Gluconolactone | G/0360 |
| Glyoxal-bis-hydroxy-anil | Di-(2-hydroxyphenylimino) ethane | D/3100 |
| Glyoxaline | Imidazole | I/0010 |
| Gold chloride | Chloroauric acid | C/4640 |
H
| Alternative Name | Catalogue Name | Ref |
| HEPES | N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid | H/2230 |
| n-Heptanol | Heptan-1-ol | H/0250 |
| Hexadecyltrimethylammonium bromide | Cetyl trimethylammonium bromide | C/3960 |
| Hexamethylenediamine | 1,6-Diaminohexane | D/0760 |
| Hexamethylenetetramine | Hexamine | H/0300 |
| Hexane-dioic acid | Adipic acid | A/1000 |
| n-Hexoic acid | n-Hexanoic acid | H/0440 |
| Hexyl alcohol | Hexan-1-ol | H/0450 |
| HFBA | Heptafl uorobutyric acid | 17280 |
| HMDS | 1,1,1,3,3,3-Hexamethyldisilazane | 12058 |
| Homidium bromide | Ethidium(R) bromide | E/P700 |
| Hydrazine salts | Hydrazinium chloride | H/0600 |
| Hydrofl uorosilicic acid | Fluorosilicic acid | F/1400 |
| Hydroquinone | Quinol | Q/0150 |
| Hydroquinone-1, 4-dihydroxybenzene | Quinol | Q/0150 |
| Hydroxyacetic acid | Glycolic acid | G/0803 |
| Hydroxylamine salts | Hydroxylammonium chloride | H/1900 |
I
| Alternative Name | Catalogue Name | Ref |
| IDA | Methylated spirits | M/4450 |
| Iminazole | Imidazole | I/0010 |
| Indanetrione hydrate | Ninhydrin | N/2158 |
| Industrial Denatured Alcohol | Methylated spirits | M/4450 |
| Industrial Methylated Spirits | Methylated spirits | M/4350, M/4450 |
| IPA | Propan-2-ol | P/7490 |
| Iron alum | Ammonium iron (III) sulfate | A/4720 |
L
| Alternative Name | Catalogue Name | Ref |
| Laevulose | D(-)-Fructose | F/1950 |
| Lead monoxide | Lead (II) oxide | L/1600 |
| tri-Lead trioxide | Lead oxide (minimum, read lead) | L/1700 |
| Litharge canary yellow | Lead (II) oxide | L/1600 |
| Lithium aluminium hydride | Aluminium lithium hydride | A/2160 |
| Lithium borate | Lithium metaborate | L/2330 |
| Luminol | 3-Aminophthalhydrazide | A/3150 |
M
| Alternative Name | Catalogue Name | Ref |
| Magenta | Fuchsin | F/P770 |
| Magnesium silicate, hydrous | Talc | T/0050 |
| Mannite | Mannitol | M/2400 |
| Mercaptoacetic acid | Thioglycolic acid | T/1000 |
| Mercury-nitric acid solution | Millon's reagent | J/5300 |
| MES | 2-(N-Morpholino)ethane-sulfonic acid | M/5760 |
| p-Methoxybenzaldehyde | Anisaldehyde | A/7490 |
| o-Methoxyphenol | Guaiacol | G/0950 |
| Methylal | Dimethoxymethane | D/3400 |
| Methyl alcohol | Methanol | M/3900 |
| p-Methylaminophenol sulfate | Metol | M/5400 |
| Methylbenzene | Toluene | T/2150 |
| 3-Methylbutan-1-ol | iso-Amyl alcohol | A/6960 |
| 2-Methylbutane | iso-Pentane | P/1030 |
| 3-Methyl-butyl acetate | iso-Amyl acetate | A/6880 |
| Methyl iso-butyl ketone | 4-Methylpentan-2-one | M/5078 |
| Methyl cellosolve | 2-Methoxyethanol | M/4100 |
| Methyl cyanide | Acetonitrile | A/0620 |
| Methylene chloride | Dichloromethane | D/1850 |
| Methylene iodide | Di-iodomethane | D/3200 |
| Methyl ethanoate | Methyl acetate | M/4150 |
| Methyl ethyl ketone | Ethyl methyl ketone | E/1450 |
| Methyl iodide | Iodomethane | I/0800 |
| Methyl phenyl ether | Anisole | A/7600 |
| 4-Methylphenylamine | p-Toluidine | T/2650 |
| Methyl phthalate | Dimethyl phthalate | D/4050 |
| 2-Methyl propan-1-ol | iso-Butyl alcohol | B/5100 |
| 2-Methyl propan-2-ol | tert-Butyl alcohol | B/5250 |
| 5-Methylresorcinol | Orcinol, monohydrate | O/0351 |
| Methyl sulfate | Dimethyl sulfate | 11682 |
| Methylthionine chloride | Methylene blue | M/4850 |
| Methylthymol blue complexone | Thymolsulfonephthalein di-(methyliminodiacetic acid) | T/1500 |
| 4-Methylumbelliferyl-B-D-glucuronide trihydrate | MUG | BPE4003 |
| Microcosmic salt | Ammonium sodium hydrogen orthophosphate | A/6280 |
| Milk sugar | Lactose | L/0200 |
| Mordant black 11 | Eriochrome black T | E/0100 |
| Mordant black 17 | Eriochrome blue-black R | E/0200 |
| 3-(4-Morpholino)propanesulfonic acid | MOPS biological buffer | BPE308 |
| Murexide | Ammonium purpurate | A/6200 |
N
| Alternative Name | Catalogue Name | Ref |
| Naphthyl bromide | 1-Bromonaphthalene | B/4550 |
| NEDD | N-1-Naphthylethylene diamine dihydrochloride | N/1250 |
| Nickel ammonium salts | Ammonium nickel (II) sulfate | A/5800 |
| NORIT | Activated charcoal | C/4120 |
O
| Alternative Name | Catalogue Name | Ref |
| Cis-Octadec-9-enoic acid | Oleic acid | O/0150 |
| iso-Octane | 2,2,4-Trimethylpentane | T/3600 |
| n-Octoic acid | n-Octanoic acid | O/0030 |
| Octylphenoxyethoxyethanol | Triton X-100 | T/3751 |
| Oil almond | Almond oil | A/1565 |
| Ottawa sand | Sand, Ottawa | S/0365 |
| Oxine | 8-Hydroxyquinoline | H/2500 |
P
| Alternative Name | Catalogue Name | Ref |
| Page blue 83 | Coomassie(R) Blue R250 | C/P540 |
| Page blue G90 | Coomassie(R) Blue G250 | C/P541 |
| PAN | 1-(2-Pyridylazo)-2-naphthol | P/8000 |
| PAR | 4-(2-Pyridylazo)-resorcinol, monosodium salt | P/8040 |
| Pentane | Petroleum ether boiling range 30 to 40°C | P/1960 |
| Pentyl acetate | n-Amyl acetate | A/6800 |
| Perchloroethylene | Tetrachloroethylene | T/0550 |
| Perfl uorobutyric acid | Heptafl uorobutyric acid | 17280 |
| Phenol reagent | Folins & Ciocalteus reagent | J/4100 |
| Phenol tris | Phenol liquefi ed washed in Tris buffer | T/P633 |
| Phenylacetone | Benzyl methyl ketone | B/2856 |
| N-Phenylethanamide | Acetanilide | A/0200 |
| Phenyl ether | Diphenyl ether | 13060 |
| Phenylethylene | Styrene | S/8480 |
| Phenyl methyl ether | Anisole | A/7600 |
| Phosphate buffered formaldehyde solution | Formaldehyde solution, phosphate buffered | F/1520 |
| Phosphomolybdic acid | dodeca-Molybdophosphoric acid | M/5650 |
| Phosphoric acid, meta | Metaphosphoric acid | M/3851 |
| Phosphoric acid, ortho | Orthophosphoric acid | O/0450 |
| Phosphoric oxide | Phosphorus pentoxide | P/2960 |
| Phosphorous oxychloride | Phosphoryl chloride | P/3280 |
| Phosphotungstic acid | dodeca-Tungstophosphoric acid | T/3902 |
| PMSF | Phenylmethylsulfonyl fl uoride | P/2805 |
| Polyoxyethylene (20) sorbitan monolaurate | Tween 20 | T/4206 |
| Polyoxyethylene (20) sorbitan monooleate | Tween 80 | T/4203 |
| Polyoxyethylene (20) sorbitan monopalmitate | Tween 40 | T/4205 |
| Polyvinyl alcohol | Vinyl alcohol, polymerised | V/0200 |
| Potassium alum | Aluminium potassium sulfate | A/2400 |
| Potassium bicarbonate | Potassium hydrogen carbonate | P/5080 |
| Potassium bisulfate | Potassium hydrogen sulfate | P/5420 |
| Potassium bisulfi te | Potassium metabisulfi te | P/5920 |
| Potassium chloroplatinite | Potassium tetrachloroplatinate (II) | P/7170 |
| Potassium ferricyanide | Potassium hexacyanoferrate (III) | P/4880 |
| Potassium ferrocyanide | Potassium hexacyanoferrate (II) | P/4920 |
| Potassium phosphate | di-Potassium hydrogen orthophosphate, Potassium dihydrogen orthophosphate | P/5240, P/4760 |
| Potassium phosphate, tribasic | tri-Potassium orthophosphate | P/6280 |
| Potassium tartrate acid | Potassium hydrogen tartrate | P/5480 |
| PPO | 2,5-Diphenyloxazole | D/4865 |
| iso-Propanol | Propan-2-ol | P/7490 |
| Propanone | Acetone | A/0520 |
| Proprionitrile | Acetonitrile | A/0620 |
| n-Propyl alcohol | Propan-1-ol | P/7480 |
| iso-Propyl alcohol | Propan-2-ol | P/7490 |
| iso-Propyl iodide | 2-Iodopropane | I/0831 |
| Propylene glycol | Propane-1,2-diol | P/7440 |
| Propylene oxide | 1,2-Epoxypropane | E/0050 |
| Propyl ethanoate | iso-Propyl acetate | P/7640 |
| Propyl ethanoate | n-Propyl acetate | P/7600 |
| iso-Propyl ether | Di-iso-propyl ether | D/5000 |
| iso-Propyl-B-D-thiogalactopyranoside | IPTG | BPE1755 |
| PVP | Polyvinyl pyrrolidone | P/3700 |
| Pyridine hydrochloride | Pyridinium chloride | P/7980 |
| Pyrocatechol | Catechol | C/3320 |
| Pyrogallic acid | Pyrogallol | P/8080 |
| Pyrogallol solution | Potassium pyrogallate solution | J/7000 |
Q
| Alternative Name | Catalogue Name | Ref |
| Quinone | p-Benzoquinone | B/2150 |
R
| Alternative Name | Catalogue Name | Ref |
| Red lead | Lead oxide | L/1700 |
| Rennin | Rennet | R/0030 |
| Rochelle salt | Potassium sodium tartrate | P/6800 |
| Rock salt | Sodium chloride | S/3100 |
| Rubeanic acid | Dithio-oxamide | D/5150 |
S
| Alternative Name | Catalogue Name | Ref |
| Saccharose | Sucrose | S/8560 |
| Salicylsulfonic acid | 5-Sulfosalicylic acid | S/8850 |
| Screened methyl orange | Methyl orange-xylene cyanol FF | M/5020L |
| Selenious acid | Selenous acid | S/0520 |
| Sequestric acid | Diaminoethanetetra-acetic acid | D/0450 |
| Silane A-174 | 3-Trimethoxysilyl-propyl-methacrylate | T/3350 |
| Silicon carbide | Carborundum | C/3090 |
| Sodium alizarin sulfonate | Alizarin Red | A/1520 |
| Sodium ammonium salts | Ammonium sodium hydrogen orthophosphate | A/6280 |
| Sodium barbitone | Barbitone sodium | B/0100 |
| Sodium bicarbonate | Sodium hydrogen carbonate | S/4200 |
| Sodium bisulfate | Sodium hydrogen sulfate | S/4690 |
| Sodium bisulfi te | Sodium metabisulfi te | S/5240 |
| Sodium borate | di-Sodium tetraborate | S/7000 |
| Sodium carboxymethyl-cellulose | Carboxymethyl cellulose sodium salt | C/3100 |
| di-Sodium dihydrogen ethylene-diamine-NNN'N'-tetra acetate | Diaminoethanetetra-acetic acid disodium salt | D/0650 |
| Sodium dimethylarsenate | Sodium cacodylate | S/2660 |
| Sodium dodecyl sulphate | Sodium dodecyl sulfate | S/5200 |
| Sodium dodecyl sulphate | Sodium dodecyl sulfate (SDS) solution | BPE1311 |
| Sodium ferrocyanide | Sodium hexacyanoferrate (II) | S/3840 |
| Sodium hydrosulfi te | Sodium dithionite | S/3800 |
| Sodium hyposulfi te | Sodium thiosulfate | S/7160 |
| Sodium lauryl sulfate | Sodium dodecyl sulfate | S/5200 |
| Sodium mercaptoacetate | Sodium thioglycolate | 14889 |
| Sodium metaperiodate | Sodium periodate | S/5980 |
| Sodium metaphosphate | Sodium hexametaphosphate | S/4120 |
| Sodium phosphate, dibasic | di-Sodium hydrogen orthophosphate | S/4360 |
| Sodium phosphate, monobasic | Sodium dihydrogen orthophosphate | S/3720 |
| Sodium phosphate, tribasic | tri-Sodium orthophosphate | S/5760 |
| Sodium potassium tartrate | Potassium sodium tartrate | P/6800 |
| Sodium rhodizonate | Rhodizonic acid sodium salt | R/0300 |
| Sodium silicofl uoride | Sodium fl uorosilicate | S/4000 |
| Sodium sulfocyanide | Sodium thiocyanate | S/7120 |
| Sodium sulphate | Sodium sulfate anhydrous | S/6600 |
| Sodium tartrate, acid | Sodium hydrogen tartrate | S/4720 |
| Sodium tetraoxodisulfate | Sodium dithionite | S/3800 |
| Solochrome black | Eriochrome black T | E/0100 |
| Solochrome dark blue | Eriochrome blue-black R | E/0200 |
| Sörensens salt | di-Sodium hydrogen orthophosphate, dihydrate | S/4440 |
| Spirit, methylated | Methylated spirits, industrial | M/4450 |
| SSC | Saline-sodium citrate, 20X solution | BPE1325 |
| SSPE | Saline-sodium phosphate-EDTA, 20X solution | BPE1328 |
| Standard water solution | Water solution, Karl Fischer | J/9000 |
| Stannic salts | Tin (IV) chloride | T/1661 |
| Stannous salts | Tin (II) chloride | T/1645 |
| Sulfur dioxide | Sulfurous acid | J/8500 |
| Sulphur dioxide gas | Sulfur dioxide gas | S/9081 |
| Sulphur hexafl uoride | Sulfur hexafl uoride gas | S/9101 |
| Sulphur standard solution | Sulfur standard solution | J/8323 |
| Sulphuric acid | Sulfuric acid | S/9120 |
| Sulphurous acid | Sulfurous acid | J/8500 |
T
| Alternative Name | Catalogue Name | Ref |
| T4 DNA Ligase | Modifying enzymes | BPET4DL-500 |
| T4 Polynucleotide Kinase | Modifying enzymes | BPET4PK-050 |
| TAE buffer | Tris-Acetate-EDTA buffer | BPE1335-1 |
| TBE buffer | Buffer concentrate TBE | T/P050 |
| TBE buffer | Tris-Borate-EDTA buffer | BPE1333 |
| TEA | Triethylamine | T/3200 |
| TEMED | NNN'N'-Tetramethyl-ethylenediamine | T/P190 |
| TES | N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic acid | T/3720 |
| Tetra-n-butyl ammonium dihydrogen phosphate | Tetra butyl ammonium phosphate | T/0505 |
| Tetrahydropyrrole | Pyrrolidine | P/8140 |
| Tetralin | Tetrahydronaphthalene | T/0850 |
| Tetrazolium blue chloride | Blue tetrazolium | B/3660 |
| Tetrazolium salt | 2,3,5-Triphenyl-tetrazolium chloride | T/3700 |
| TFA | Trifl uoroacetic acid | T/3255 |
| THF | Tetrahydrofuran | T/0700 |
| Thiocarbamide | Thiourea | T/1150 |
| Thiodiglycol | 2,2'-Thiodiethanol | T/0998 |
| Tiron | 1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt | D/3000 |
| Titanium (IV) isopropoxide | Tetra-iso-propyl orthotitanate | 1470 |
| TMCS | Trimethylchlorosilane | T/3520 |
| TMSI | N-Trimethylsilylimidazole | T/3622 |
| Toluene-4-sulfonchloride | Toluene-4-sulfonyl chloride | T/2550 |
| Tosyl chloride | Toluene-4-sulfonyl chloride | T/2550 |
| Trichloromethane | Chloroform | C/4920 |
| Tricine | N-Tris(hydroxymethyl) methyl glycine | T/P505 |
| Triethylene glycol | Trigol | T/3270 |
| 1,2,3-Trihydroxybenzene | Pyrogallol | P/8080 |
| Triketohydrindene hydrate | Ninhydrin | N/2158 |
| 1,3,5-Trimethylbenzene | Mesitylene | 12558 |
| Trimethylene glycol | Propane-1,3-diol | P/7450 |
| 2,4,6-Trimethylpyridine | 2,4,6-Collidine | C/6940 |
| Tris buffer | Tris (hydroxymethyl)aminomethane | T/3710 |
| Tris washed-phenol | Phenol liquefi ed | T/P633 |
| Tritolyl phosphate | Tri-aryl phosphate | T/2820 |
V
| Alternative Name | Catalogue Name | Ref |
| Vitamin C | L-Ascorbic acid | A/8880 |
W
| Alternative Name | Catalogue Name | Ref |
| Wanklyn's soap solution | Soap solution, Wanklyn | J/7500 |
| White spirit | Turpentine substitute | T/4200 |