accessibility menu, dialog, popup

UserName

CAS RN 998-29-8

CH3SiH3CCH3

CAS RN 998-29-8

IUPAC Name: tripropylsilyl
Synonyms: tripropyl silane
Molecular Weight (g/mol): 157.35
Molecular Formula: C9H21Si
InChi Key: MMYRBBZVCDXGHG-UHFFFAOYSA-N
SMILES: CCC[Si](CCC)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results