CAS RN 99300-68-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydroxy Torsemide, TRC
CAS: 99300-68-2 Molecular Formula: C16H20N4O4S Molecular Weight (g/mol): 364.42 Synonym: 4-[[3-(Hydroxymethyl)phenyl]amino]-N-[[(1-methylethyl)amino]carbonyl]-3-pyridinesulfonamide,AC 3534 IUPAC Name: 1-[4-[3-(hydroxymethyl)anilino]pyridin-3-yl]sulfonyl-3-propan-2-ylurea SMILES: CC(C)NC(=O)NS(=O)(=O)C1=C(C=CN=C1)NC2=CC=CC(=C2)CO