accessibility menu, dialog, popup

UserName

CH3OHNOOH3C

CAS RN 98081-83-5

IUPAC Name: methyl 6-methoxy-1H-indole-2-carboxylate
Synonyms: methyl 6-methoxy-1H-indole-2-carboxylate
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: OPUUCOLVBDQWEY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=CC=C(OC)C=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results