CAS RN 96315-26-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl 2-(Chloromethyl)oxazole-5-carboxylate (~90%), TRC
CAS: 96315-26-3 Molecular Formula: C7H8ClNO3 Molecular Weight (g/mol): 189.6 Synonym: 2-(Chloromethyl)-5-oxazolecarboxylic Acid Ethyl Ester,2-Chloromethyl-5-ethoxycarbonyloxazole IUPAC Name: ethyl 2-(chloromethyl)-1,3-oxazole-5-carboxylate SMILES: CCOC(=O)c1oc(CCl)nc1