CAS RN 95-02-3
CAS RN 95-02-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Amino-5-aminomethyl-2-methylpyrimidine, TRC
CAS: 95-02-3 Molecular Formula: C6 H10 N4 Molecular Weight (g/mol): 138.17 Synonym: 5-Pyrimidinemethanamine, 4-amino-2-methyl-;Pyrimidine, 4-amino-5-(aminomethyl)-2-methyl- (6CI,8CI),4-Amino-2-methyl-5-pyrimidinemethanamine,2-Methyl-4-amino-5-(aminomethyl)pyrimidine,4-Amino-5-aminomethyl-2-methylpyrimidine,5-(Aminomethyl)-4-amino-2-methylpyrimidine,Grewe diamine,5-(Aminomethyl)-2-methylpyrimidin-4-amine IUPAC Name: 5-(aminomethyl)-2-methylpyrimidin-4-amine SMILES: Cc1ncc(CN)c(N)n1