accessibility menu, dialog, popup

UserName

CAS RN 942615-32-9

H3CBrCH3

CAS RN 942615-32-9

IUPAC Name: 4-bromo-9,9-dimethyl-9H-fluorene
Synonyms: 4-bromo-9,9-dimethylfluorene
Molecular Weight (g/mol): 273.17
Molecular Formula: C15H13Br
InChi Key: SXOUNESKHJIXNK-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C1C=CC=C2Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)