accessibility menu, dialog, popup

UserName

H3COOHONHOSCH3CH3CH3SOOO

CAS RN 94055-76-2

IUPAC Name: (2-{[4-(3-ethoxy-2-hydroxypropoxy)phenyl]carbamoyl}ethyl)dimethylsulfanium 4-methylbenzene-1-sulfonate
Synonyms: suplatast tosilate 甲磺司特
Molecular Weight (g/mol): 499.64
Molecular Formula: C23H33NO7S2
InChi Key: RYVJQEZJUFRANT-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S([O-])(=O)=O.CCOCC(O)COC1=CC=C(NC(=O)CC[S+](C)C)C=C1

 No Refinements