accessibility menu, dialog, popup

UserName

CAS RN 93680-80-9

H3COOO

CAS RN 93680-80-9

IUPAC Name: 1-benzofuran-3-yl acetate
Synonyms: 1-benzofuran-3-yl acetate
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: XCBLZAJCKKRBED-UHFFFAOYSA-N
SMILES: CC(=O)OC1=COC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results