accessibility menu, dialog, popup

UserName

CAS RN 936693-22-0

BrNF

CAS RN 936693-22-0

IUPAC Name: 2-(2-bromo-6-fluorophenyl)acetonitrile
Synonyms: 2-(2-bromo-6-fluorophenyl)acetonitrile
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H5BrFN
InChi Key: QFEXNLSISKHIBO-UHFFFAOYSA-N
SMILES: FC1=C(CC#N)C(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results