accessibility menu, dialog, popup

UserName

CAS RN 936250-21-4

BOOH3CCH3H3CH3CNNNOOCH3CH3CH3

CAS RN 936250-21-4

IUPAC Name: tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine-1-carboxylate
Molecular Weight (g/mol): 389.3
Molecular Formula: C20H32BN3O4
InChi Key: NQBNJXYYAKBREJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=NC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results