accessibility menu, dialog, popup

UserName

CH3OFFF

CAS RN 93249-45-7

IUPAC Name: 4-methyl-3-(trifluoromethyl)benzaldehyde
Synonyms: 4-methyl-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 188.15
Molecular Formula: C9H7F3O
InChi Key: FRLOLHJJROPPKU-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=O)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results