accessibility menu, dialog, popup

UserName

CAS RN 92814-00-1

CH3FFBrFF

CAS RN 92814-00-1

IUPAC Name: 1-(bromomethyl)-2,3,5,6-tetrafluoro-4-methylbenzene
Synonyms: 1-(bromomethyl)-2,3,5,6-tetrafluoro-4-methylbenzene
Molecular Weight (g/mol): 257.03
Molecular Formula: C8H5BrF4
InChi Key: JGBVMYAEIPKDQB-UHFFFAOYSA-N
SMILES: CC1=C(F)C(F)=C(CBr)C(F)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results