CAS RN 92414-09-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(1RS)-1-[(Dimethylamino)methyl]-1-phenylpropyl 3,4,5-Trimethoxybenzoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
1-(Dimethylamino)-2-phenyl-2-butanyl 3,4,5-trimethoxybenzoate, TRC
CAS: 92414-09-0 Molecular Formula: C22 H29 N O5 Molecular Weight (g/mol): 387.47 Synonym: (1RS)-1-[(Dimethylamino)methyl]-1-phenylpropyl 3,4,5-Trimethoxybenzoate,Trimebutine Imp. D (EP) IUPAC Name: [1-(dimethylamino)-2-phenylbutan-2-yl] 3,4,5-trimethoxybenzoate SMILES: CCC(CN(C)C)(OC(=O)c1cc(OC)c(OC)c(OC)c1)c2ccccc2