CAS RN 915923-65-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(5-Formylpyrimidin-2-yl)glycine, TRC
CAS: 915923-65-8 Molecular Formula: C7H7N3O3 Molecular Weight (g/mol): 181.14 SMILES: OC(=O)CNc1ncc(C=O)cn1