accessibility menu, dialog, popup

UserName

CAS RN 90390-33-3

H2NFCl

CAS RN 90390-33-3

IUPAC Name: 1-(3-chloro-5-fluorophenyl)methanamine
Synonyms: 1-(3-chloro-5-fluorophenyl)methanamine
Molecular Weight (g/mol): 159.59
Molecular Formula: C7H7ClFN
InChi Key: XJCYOVBALKWQQC-UHFFFAOYSA-N
SMILES: NCC1=CC(F)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results