CAS RN 902836-14-0
CAS RN 902836-14-0
IUPAC Name:
2-[4-(ethoxymethyl)phenyl]acetic acid
Synonyms:
[4-(ethoxymethyl)phenyl]acetic acid
Molecular Weight (g/mol):
194.23
Molecular Formula:
C11H14O3
InChi Key:
JSRDATKLAHCUBB-UHFFFAOYSA-N
SMILES:
CCOCC1=CC=C(CC(O)=O)C=C1
1
–
1
of
1
results
1
4-(Ethoxymethyl)phenylacetic acid, Thermo Scientific™
CAS: 902836-14-0 Molecular Formula: C11H14O3 Molecular Weight (g/mol): 194.23 MDL Number: MFCD08061030 InChI Key: JSRDATKLAHCUBB-UHFFFAOYSA-N Synonym: 4-ethoxymethyl phenyl acetic acid,4-ethoxymethyl phenylacetic acid,2-4-ethoxymethyl phenyl acetic acid,4-ethoxymethyl-phenyl-acetic acid PubChem CID: 24208841 IUPAC Name: 2-[4-(ethoxymethyl)phenyl]acetic acid SMILES: CCOCC1=CC=C(C=C1)CC(=O)O