CAS RN 9000-59-3
CAS RN 9000-59-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Shellac Orange, Pure, Flake, Fisher Chemical™
CAS: 9000-59-3 MDL Number: 148309