CAS RN 897598-47-9
CAS RN 897598-47-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(4-Fluoro-2-nitrophenyl)azepane, 97%, Thermo Scientific™
CAS: 897598-47-9 Synonym: 1-(4-Fluoro-2-nitrophenyl)hexamethyleneimine