accessibility menu, dialog, popup

UserName

H3COOFFFOH

CAS RN 895572-36-8

IUPAC Name: 2-methoxy-5-(trifluoromethoxy)phenol
Synonyms: 2-methoxy-5-(trifluoromethoxy)phenol
Molecular Weight (g/mol): 208.14
Molecular Formula: C8H7F3O3
InChi Key: PORCXQLOOCXXPL-UHFFFAOYSA-N
SMILES: COC1=C(O)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results