accessibility menu, dialog, popup

UserName

CAS RN 89466-05-7

BNOONH2HOOH

CAS RN 89466-05-7

IUPAC Name: (3-amino-5-nitrophenyl)boronic acid
Synonyms: 3-amino-5-nitrophenylboronic acid
Molecular Weight (g/mol): 181.94
Molecular Formula: C6H7BN2O4
InChi Key: KLVJZQJSYAKUKK-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1)B(O)O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results