accessibility menu, dialog, popup

UserName

CAS RN 886500-31-8

ClOOHFFF

CAS RN 886500-31-8

IUPAC Name: 2-[2-chloro-6-(trifluoromethyl)phenyl]acetic acid
Synonyms: [2-chloro-6-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: QVSWFQSUELDCNS-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(C=CC=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results