accessibility menu, dialog, popup

UserName

CAS RN 886499-17-8

CH3OFBrF

CAS RN 886499-17-8

IUPAC Name: 1-(bromomethyl)-2,4-difluoro-3-methoxybenzene
Synonyms: 1-(bromomethyl)-2,4-difluoro-3-methoxybenzene
Molecular Weight (g/mol): 237.04
Molecular Formula: C8H7BrF2O
InChi Key: DXDNJSHUSXGESC-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(CBr)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results