accessibility menu, dialog, popup

UserName

NH2HNFFFFFF

CAS RN 886-35-1

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]hydrazine
Synonyms: [3,5-bis(trifluoromethyl)phenyl]hydrazine
Molecular Weight (g/mol): 244.14
Molecular Formula: C8H6F6N2
InChi Key: OULWACKZBGBRNT-UHFFFAOYSA-N
SMILES: NNC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results