accessibility menu, dialog, popup

UserName

CAS RN 885952-12-5

CH3NIH3CNH2

CAS RN 885952-12-5

IUPAC Name: 5-iodo-4,6-dimethylpyridin-2-amine
Synonyms: 5-iodo-4,6-dimethylpyridin-2-amine
Molecular Weight (g/mol): 248.07
Molecular Formula: C7H9IN2
InChi Key: KJDKMUXRVKLPJF-UHFFFAOYSA-N
SMILES: CC1=CC(N)=NC(C)=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results