CAS RN 885703-64-0
CAS RN 885703-64-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
c-Fms-IN-1, MedChemExpress
MedChemExpress c-Fms-IN-1 is a FMS kinase inhibitor with an IC50 of 0.0008 μM.