accessibility menu, dialog, popup

UserName

H3COSOONHOHO

CAS RN 885269-51-2

IUPAC Name: 3-{4'-methoxy-[1,1'-biphenyl]-4-sulfonamido}propanoic acid
Synonyms: 3-{4'-methoxy-[1,1'-biphenyl]-4-sulfonamido}propanoic acid
Molecular Weight (g/mol): 335.37
Molecular Formula: C16H17NO5S
InChi Key: SDSCQHVGKWYOCF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=C(C=C1)S(=O)(=O)NCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)