CAS RN 885269-48-7
IUPAC Name:
1-{4'-methoxy-[1,1'-biphenyl]-4-sulfonamido}cyclohexane-1-carboxylic acid
Synonyms:
1-{4'-methoxy-[1,1'-biphenyl]-4-sulfonamido}cyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
389.47
Molecular Formula:
C20H23NO5S
InChi Key:
JAKSOMBIPMPTDM-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C1=CC=C(C=C1)S(=O)(=O)NC1(CCCCC1)C(O)=O
1
–
1
of
1
results
1
1-(4'-Methoxy-4-biphenylylsulfonylamino)cyclohexanecarboxylic acid, 96%, Thermo Scientific™
CAS: 885269-48-7 Molecular Formula: C20H23NO5S Molecular Weight (g/mol): 389.47 MDL Number: MFCD06409463 InChI Key: JAKSOMBIPMPTDM-UHFFFAOYSA-N Synonym: 1-4'-methoxy-1,1'-biphenyl-4-sulfonamido cyclohexane-1-carboxylic acid,1-4'-methoxy-1,1'-biphenyl-4-ylsulfonamido cyclohexanecarboxylic acid,1-4'-methoxy-4-biphenylylsulfonylamino cyclohexanecarboxylic acid,1-4'-methoxy 1,1'-biphenyl-4-sulfonyl amino cyclohexane-1-carboxylic acid PubChem CID: 44669172 IUPAC Name: 1-[[4-(4-methoxyphenyl)phenyl]sulfonylamino]cyclohexane-1-carboxylic acid SMILES: COC1=CC=C(C=C1)C1=CC=C(C=C1)S(=O)(=O)NC1(CCCCC1)C(O)=O