accessibility menu, dialog, popup

UserName

CH3OOFFF

CAS RN 883541-11-5

IUPAC Name: 4-ethoxy-3-(trifluoromethyl)benzaldehyde
Synonyms: 4-ethoxy-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 218.18
Molecular Formula: C10H9F3O2
InChi Key: NGQLIMKJXPGLBN-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(C=O)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results