accessibility menu, dialog, popup

UserName

CAS RN 877399-50-3

CH3H3CCH3OONNNBr

CAS RN 877399-50-3

IUPAC Name: tert-butyl 4-(4-bromo-1H-pyrazol-1-yl)piperidine-1-carboxylate
Synonyms: tert-butyl 4-(4-bromopyrazol-1-yl)piperidine-1-carboxylate
Molecular Weight (g/mol): 330.23
Molecular Formula: C13H20BrN3O2
InChi Key: IYNZAVDBHAQODX-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(Br)C=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results