accessibility menu, dialog, popup

UserName

CAS RN 871329-77-0

BSOONHCH3H3CHOOH

CAS RN 871329-77-0

IUPAC Name: {3-[(propan-2-yl)sulfamoyl]phenyl}boronic acid
Synonyms: 3-(isopropylsulfamoyl)phenylboronic acid
Molecular Weight (g/mol): 243.08
Molecular Formula: C9H14BNO4S
InChi Key: KCJMBNYQVZPTRG-UHFFFAOYSA-N
SMILES: CC(C)NS(=O)(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results