accessibility menu, dialog, popup

UserName

CH3NBrF

CAS RN 864830-16-0

IUPAC Name: 5-bromo-2-fluoro-4-methylpyridine
Synonyms: 5-bromo-2-fluoro-4-methylpyridine
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: PFODFMZCBQUYNG-UHFFFAOYSA-N
SMILES: CC1=CC(F)=NC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results