CAS RN 864685-26-7
CAS RN 864685-26-7
IUPAC Name:
tert-butyl 5-cyano-3-iodo-1H-indole-1-carboxylate
Synonyms:
tert-butyl 5-cyano-3-iodoindole-1-carboxylate
Molecular Weight (g/mol):
368.17
Molecular Formula:
C14H13IN2O2
InChi Key:
GADLXNFRAAUWAD-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)N1C=C(I)C2=CC(=CC=C12)C#N
1
–
1
of
1
results
1
1-Boc-3-iodoindole-5-carbonitrile, 97%, Thermo Scientific™
CAS: 864685-26-7 Molecular Formula: C14H13IN2O2 Molecular Weight (g/mol): 368.17 MDL Number: MFCD05864767 InChI Key: GADLXNFRAAUWAD-UHFFFAOYSA-N Synonym: tert-butyl 5-cyano-3-iodo-1h-indole-1-carboxylate,1-boc-5-cyano-3-iodo-indole,1-boc-5-cyano-3-iodoindole,5-cyano-3-iodoindole-1-carboxylic acid tert-butyl ester,5-cyano-3-iodo-1h-indole-1-carboxylic acid tert-butyl ester,pubchem7950,1-tert-butoxycarbonyl-5-cyano-3-iodoindole,tert-butyl 5-cyano-3-iodo-indole-1-carboxylate,tert-butyl 5-cyano-3-iodanyl-indole-1-carboxylate,tert-butyl5-cyano-3-iodo-1h-indole-1-carboxylate PubChem CID: 17749979 SMILES: CC(C)(C)OC(=O)N1C=C(I)C2=CC(=CC=C12)C#N