CAS RN 85326-06-3
CAS RN 85326-06-3
IUPAC Name:
2-amino-9-[5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
Synonyms:
2-amino-9-[5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Molecular Weight (g/mol):
251.25
Molecular Formula:
C10H13N5O3
InChi Key:
OCLZPNCLRLDXJC-UHFFFAOYNA-N
SMILES:
NC1=NC(=O)C2=C(N1)N(C=N2)C1CCC(CO)O1
1
–
1
of
1
results
1
2',3'-Dideoxyguanosine, 90+%, Thermo Scientific™
CAS: 85326-06-3 Molecular Formula: C10H13N5O3 Molecular Weight (g/mol): 251.25 MDL Number: MFCD00057074 InChI Key: OCLZPNCLRLDXJC-NTSWFWBYSA-N Synonym: 2',3'-dideoxyguanosine,guanosine, 2',3'-dideoxy,2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one,dideoxyguanosine,2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one,deoxy-deoxy-guanosine,2,3-didioxyguanosine,2', 3'-dideoxyguanosine,2,3-dideoxyguanosine,chembl78101 PubChem CID: 64974 IUPAC Name: 2-amino-9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one SMILES: NC1=NC2=C(N=CN2[C@H]2CC[C@@H](CO)O2)C(=O)N1