accessibility menu, dialog, popup

UserName

CAS RN 851263-13-3

CH3OOH3CFFF

CAS RN 851263-13-3

IUPAC Name: 1-[4-ethoxy-3-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[4-ethoxy-3-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 232.2
Molecular Formula: C11H11F3O2
InChi Key: KWNZFVILRICPQN-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(C=C1)C(C)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results